missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]aniline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1202-00-2
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
InChI Key OHDPNHWSANKRNF-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy aniline, 2-2-dimethylamino-ethoxy-phenylamine, benzenamine,2-2-dimethylamino ethoxy, benzenamine, 2-2-dimethylamino ethoxy, 2-2-aminophenoxy ethyl dimethylamine, 2-2-dimethylaminoethyloxy aniline
PubChem CID 6484656
IUPAC Name 2-[2-(dimethylamino)ethoxy]aniline
SMILES CN(C)CCOC1=CC=CC=C1N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC51314CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC51314DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC51314DE
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1202-00-2
180.251
2-2-dimethylamino ethoxy aniline, 2-2-dimethylamino-ethoxy-phenylamine, benzenamine,2-2-dimethylamino ethoxy, benzenamine, 2-2-dimethylamino ethoxy, 2-2-aminophenoxy ethyl dimethylamine, 2-2-dimethylaminoethyloxy aniline
2-[2-(dimethylamino)ethoxy]aniline
C10H16N2O
OHDPNHWSANKRNF-UHFFFAOYSA-N
6484656
CN(C)CCOC1=CC=CC=C1N

Specifications

1202-00-2
C10H16N2O
OHDPNHWSANKRNF-UHFFFAOYSA-N
2-[2-(dimethylamino)ethoxy]aniline
6484656
97%
97%
2-2-dimethylamino ethoxy aniline, 2-2-dimethylamino-ethoxy-phenylamine, benzenamine,2-2-dimethylamino ethoxy, benzenamine, 2-2-dimethylamino ethoxy, 2-2-aminophenoxy ethyl dimethylamine, 2-2-dimethylaminoethyloxy aniline
CN(C)CCOC1=CC=CC=C1N
180.251
180.25
2-[2-(Dimethylamino)ethoxy]aniline
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents