accessibility menu, dialog, popup

UserName

H3CNCH3OH2N

CAS RN 1202-00-2

IUPAC Name: 2-[2-(dimethylamino)ethoxy]aniline
Synonyms: 2-[2-(dimethylamino)ethoxy]aniline
Molecular Weight (g/mol): 180.25
Molecular Formula: C10H16N2O
InChi Key: OHDPNHWSANKRNF-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)