missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-1,3-benzodioxole 98.0+%, TCI America™
$199.46 - $546.17
Chemical Identifiers
| CAS | 14005-14-2 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD08061594 |
| InChI Key | BWBIFYYKIWPTRV-UHFFFAOYSA-N |
| Synonym | 1,2-(Isopropylidenedioxy)benzene |
| PubChem CID | 285869 |
| IUPAC Name | 2,2-dimethyl-1,3-benzodioxole |
| SMILES | CC1(OC2=CC=CC=C2O1)C |
Chemical Identifiers
| 14005-14-2 | |
| 150.177 | |
| BWBIFYYKIWPTRV-UHFFFAOYSA-N | |
| 285869 | |
| CC1(OC2=CC=CC=C2O1)C |
| C9H10O2 | |
| MFCD08061594 | |
| 1,2-(Isopropylidenedioxy)benzene | |
| 2,2-dimethyl-1,3-benzodioxole |
Specifications
| 14005-14-2 | |
| Yellow | |
| C9H10O2 | |
| 5 g | |
| BWBIFYYKIWPTRV-UHFFFAOYSA-N | |
| 2,2-dimethyl-1,3-benzodioxole | |
| 285869 | |
| ≥98.0% (GC) | |
| 2,2-Dimethyl-1,3-benzodioxole |
| 3°C | |
| 182°C | |
| MFCD08061594 | |
| 1,2-(Isopropylidenedioxy)benzene | |
| CC1(OC2=CC=CC=C2O1)C | |
| 150.177 | |
| 150.18 | |
| Liquid |
Safety and Handling
TSCA : No