accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 14005-14-2

H3CH3COO

CAS RN 14005-14-2

IUPAC Name: 2,2-dimethyl-2H-1,3-benzodioxole
Synonyms: 2,2-dimethyl-1,3-benzodioxole
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: BWBIFYYKIWPTRV-UHFFFAOYSA-N
SMILES: CC1(C)OC2=CC=CC=C2O1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.