missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxypropane 98.0+%, TCI America™

Catalog No. A0057500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0057500ML 500 mL
A005725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0057500ML Supplier TCI America Supplier No. A0057500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-76-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00008479
InChI Key HEWZVZIVELJPQZ-UHFFFAOYSA-N
Synonym acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
PubChem CID 6495
IUPAC Name 2,2-dimethoxypropane
SMILES CC(C)(OC)OC

Specifications

Color Colorless
Boiling Point 80°C
Quantity 500 mL
UN Number 1993
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethoxypropane
EINECSNumber (2)-0495
TSCA Yes