accessibility menu, dialog, popup

UserName

CAS RN 77-76-9

IUPAC Name: 2,2-dimethoxypropane
Synonyms: 2,2-dimethoxypropane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: HEWZVZIVELJPQZ-UHFFFAOYSA-N
SMILES: COC(C)(C)OC
Boiling Point 
  • (3)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (5)
  • (4)
  • (2)
  • (3)
  • (4)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)