missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxypropane, 98+%

CAS: 77-76-9 | C5H12O2 | 104.15 g/mol

$113.28 - $223.09

Chemical Identifiers

CAS 77-76-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
InChI Key HEWZVZIVELJPQZ-UHFFFAOYSA-N
Synonym acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
PubChem CID 6495
IUPAC Name 2,2-dimethoxypropane
SMILES CC(C)(OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115635000
SDS View Documents
Thermo Scientific Chemicals
115635000
500 mL Glass bottle
Each for $113.28
Only null left
 
AC115630010
SDS View Documents
Thermo Scientific Chemicals
115630010
1 L Glass bottle
Each for $223.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Chemical Identifiers

77-76-9
104.15
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
2,2-dimethoxypropane
C5H12O2
HEWZVZIVELJPQZ-UHFFFAOYSA-N
6495
CC(C)(OC)OC

Specifications

77-76-9
100.0
Colorless
83.0°C
Authentic
Glass bottle
1.3760 to 1.3780
500 mL
01,268; 05,226; 13,199; 15,138
acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
HEWZVZIVELJPQZ-UHFFFAOYSA-N
2,2-dimethoxypropane
6495
104.15
Liquid
98.0
-47.0°C
0.8400g/mL
−5°C
98% min. (GC)
C5H12O2
(CH3)2C(OCH3)2
01,648
0.84
Solubility in water: 18 g/100g (25°C). Other solubilities: soluble in acetone, benzene, ccl4, ether,, n-heptane and methanol
CC(C)(OC)OC
104.15
0.452 mPa.s (25°C)
98+%
2,2-Dimethoxypropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-056-

SDS
Documents

Documents

RUO – Research Use Only