Learn More
2,2-Dimethoxypropane, 98%
CAS: 77-76-9 | C5H12O2 | 104.149 g/mol
Supplier: Thermo Scientific Chemicals A13810AE
Description
2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Dimethoxypropane acts as a dehydrating agent. It also serves as an intermediate in the synthesis of vitamin E, vitamin A and various carotenoids such as astaxanthin. It is used as a reagent for the preparation of 1,2-diols, acetonides, isopropylidene derivatives of sugars, nucleosides, methyl esters of amino acids and enol ethers.
Solubility
Soluble in benzene, carbon tetrachloride, ethyl ether and n-butane, methanol. Moderately soluble in water.
Notes
Incompatible with oxidizing agents and acids.
Specifications
| 2,2-Dimethoxypropane | |
| -47°C | |
| 79°C to 81°C | |
| 98% | |
| C5H12O2 | |
| MFCD00008479 | |
| UN3271 | |
| acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan | |
| HEWZVZIVELJPQZ-UHFFFAOYSA-N | |
| 2,2-dimethoxypropane | |
| 6495 | |
| 98% |
| 77-76-9 | |
| 0.848 | |
| −11°C (12°F) | |
| Sweet | |
| 1.378 | |
| 100 mL | |
| 635678 | |
| Soluble in benzene,carbon tetrachloride,ethyl ether and n-butane,methanol. Moderately soluble in water. | |
| CC(C)(OC)OC | |
| 104.149 | |
| 104.15 | |
| Liquid |
Chemical Identifiers
| 77-76-9 | |
| 104.149 | |
| HEWZVZIVELJPQZ-UHFFFAOYSA-N | |
| 6495 | |
| CC(C)(OC)OC |
| C5H12O2 | |
| MFCD00008479 | |
| acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan | |
| 2,2-dimethoxypropane |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 201-056-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only