missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dimethoxybiphenyl, 97%

Catalog No. AAB2341106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2341106 5 g
AAB2341114 25 g
AAB2341122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2341106 Supplier Thermo Scientific Chemicals Supplier No. B2341106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4877-93-4 | C14H14O2 | 214.26 g/mol

2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 4877-93-4
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00067073
InChI Key VGMKUVCDINAAFC-UHFFFAOYSA-N
PubChem CID 611816
IUPAC Name 1-methoxy-2-(2-methoxyphenyl)benzene
SMILES COC1=CC=CC=C1C1=CC=CC=C1OC

Specifications

Melting Point 152°C to 156°C
Quantity 5 g
Synonym 2,2'-dimethoxybiphenyl, 2,2'-dimethoxy-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dimethoxy, 1-methoxy-2-2-methoxyphenyl benzene, 2,2'-dimethoxy-biphenyl, biphenyl, 2,2'-dimethoxy, 2,2/'-dimethoxybiphenyl, acmc-1ao09, 1,1'-biphenyl,2,2'-dimethoxy, 2,2'-dimethoxy-1,1'-biphenyl #
Solubility Information Soluble in chloroform,methanol.
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material 2,2'-Dimethoxybiphenyl
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only