missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diisobutyl-1,3-propanediol 98.0+%, TCI America™
Supplier: TCI America D277725G
Specifications
| 2,2-Diisobutyl-1,3-propanediol | |
| 77°C | |
| 160°C | |
| MFCD00986394 | |
| 2,2-Dihydroxy-2,2-diisobutylpropane | |
| CC(C)CC(CC(C)C)(CO)CO | |
| 188.311 | |
| 188.31 | |
| Crystalline Powder |
| 10547-96-3 | |
| White | |
| C11H24O2 | |
| 25 g | |
| PTRCHMOHGGDNIJ-UHFFFAOYSA-N | |
| 2,2-bis(2-methylpropyl)propane-1,3-diol | |
| 3985625 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 10547-96-3 | |
| 188.311 | |
| PTRCHMOHGGDNIJ-UHFFFAOYSA-N | |
| 3985625 | |
| CC(C)CC(CC(C)C)(CO)CO |
| C11H24O2 | |
| MFCD00986394 | |
| 2,2-Dihydroxy-2,2-diisobutylpropane | |
| 2,2-bis(2-methylpropyl)propane-1,3-diol |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : No