missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diisobutyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7127493
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27775G 5 g
D277725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27775G Supplier TCI America Supplier No. D27775G
Only null left

Chemical Identifiers

CAS 10547-96-3
Molecular Formula C11H24O2
Molecular Weight (g/mol) 188.311
MDL Number MFCD00986394
InChI Key PTRCHMOHGGDNIJ-UHFFFAOYSA-N
Synonym 2,2-Dihydroxy-2,2-diisobutylpropane
PubChem CID 3985625
IUPAC Name 2,2-bis(2-methylpropyl)propane-1,3-diol
SMILES CC(C)CC(CC(C)C)(CO)CO

Specifications

Melting Point 77°C
Color White
Boiling Point 160°C
Quantity 5 g
Formula Weight 188.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Diisobutyl-1,3-propanediol
EINECSNumber (2)-0240
TSCA No