missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dihydroxydiphenylmethane 99.0+%, TCI America™
$188.27 - $579.71
Chemical Identifiers
| CAS | 2467-02-9 |
|---|---|
| Molecular Formula | C13H12O2 |
| Molecular Weight (g/mol) | 200.24 |
| MDL Number | MFCD00002243 |
| InChI Key | MQCPOLNSJCWPGT-UHFFFAOYSA-N |
| Synonym | 2,2'-methylenediphenol, bis 2-hydroxyphenyl methane, 2,2'-dihydroxydiphenylmethane, 2,2'-bisphenol f, 2,2'-methylenebis phenol, phenol, 2,2'-methylenebis, o-o-hydroxybenzyl phenol, 2-2-hydroxyphenyl methyl phenol, phenol, 2,2'-methylenedi, 2,2'-bis hydroxyphenyl methane |
| PubChem CID | 75575 |
| ChEBI | CHEBI:34207 |
| IUPAC Name | 2-[(2-hydroxyphenyl)methyl]phenol |
| SMILES | C1=CC=C(C(=C1)CC2=CC=CC=C2O)O |
Chemical Identifiers
| 2467-02-9 | |
| 200.24 | |
| MQCPOLNSJCWPGT-UHFFFAOYSA-N | |
| 75575 | |
| 2-[(2-hydroxyphenyl)methyl]phenol |
| C13H12O2 | |
| MFCD00002243 | |
| 2,2'-methylenediphenol, bis 2-hydroxyphenyl methane, 2,2'-dihydroxydiphenylmethane, 2,2'-bisphenol f, 2,2'-methylenebis phenol, phenol, 2,2'-methylenebis, o-o-hydroxybenzyl phenol, 2-2-hydroxyphenyl methyl phenol, phenol, 2,2'-methylenedi, 2,2'-bis hydroxyphenyl methane | |
| CHEBI:34207 | |
| C1=CC=C(C(=C1)CC2=CC=CC=C2O)O |
Specifications
| 2467-02-9 | |
| White-Yellow | |
| MFCD00002243 | |
| 2,2'-methylenediphenol, bis 2-hydroxyphenyl methane, 2,2'-dihydroxydiphenylmethane, 2,2'-bisphenol f, 2,2'-methylenebis phenol, phenol, 2,2'-methylenebis, o-o-hydroxybenzyl phenol, 2-2-hydroxyphenyl methyl phenol, phenol, 2,2'-methylenedi, 2,2'-bis hydroxyphenyl methane | |
| C1=CC=C(C(=C1)CC2=CC=CC=C2O)O | |
| 200.24 | |
| CHEBI:34207 | |
| ≥99.0% (GC) | |
| 2,2′-Dihydroxydiphenylmethane |
| 118°C | |
| C13H12O2 | |
| 1 g | |
| MQCPOLNSJCWPGT-UHFFFAOYSA-N | |
| 2-[(2-hydroxyphenyl)methyl]phenol | |
| 75575 | |
| 200.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes