missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dihydroxybiphenyl 98.0+%, TCI America™

Catalog No. p-7118685
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B081325G 25 g
B0813100G 100 g
B0813500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B081325G Supplier TCI America Supplier No. B081325G
Only null left

Chemical Identifiers

CAS 1806-29-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002210
InChI Key IMHDGJOMLMDPJN-UHFFFAOYSA-N
Synonym 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
PubChem CID 15731
ChEBI CHEBI:28970
IUPAC Name [1,1'-biphenyl]-2,2'-diol
SMILES OC1=CC=CC=C1C1=CC=CC=C1O

Specifications

Melting Point 107°C
Boiling Point 325°C
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxybiphenyl
RTECSNumber DV4200000
TSCA Yes