Learn More
2,2'-Dihydroxybiphenyl, 99%
CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol
Supplier: Thermo Scientific Chemicals A1480022
Description
2,2'-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
| 2,2'-Dihydroxybiphenyl | |
| 107°C to 111°C | |
| 314°C to 316°C | |
| C12H10O2 | |
| 100 g | |
| 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl | |
| IMHDGJOMLMDPJN-UHFFFAOYSA-N | |
| 2-(2-hydroxyphenyl)phenol | |
| 15731 | |
| 186.21 |
| 1806-29-7 | |
| 1.34 | |
| 154°C (309°F) | |
| MFCD00002210 | |
| 1638363 | |
| Insoluble in water. | |
| OC1=CC=CC=C1C1=CC=CC=C1O | |
| 186.21 | |
| CHEBI:28970 | |
| 99% |
Chemical Identifiers
| 1806-29-7 | |
| 186.21 | |
| IMHDGJOMLMDPJN-UHFFFAOYSA-N | |
| 15731 | |
| OC1=CC=CC=C1C1=CC=CC=C1O |
| C12H10O2 | |
| MFCD00002210 | |
| 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl | |
| CHEBI:28970 |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318
EINECSNumber : 217-303-0
RTECSNumber : DV4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only