missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dihydroxybiphenyl, 99%

Catalog No. AAA1480036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1480036 500 g
AAA1480014 25 g
AAA1480022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1480036 Supplier Thermo Scientific Chemicals Supplier No. A1480036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol

2,2'-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dihydroxybiphenyl, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 1806-29-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002210
InChI Key IMHDGJOMLMDPJN-UHFFFAOYSA-N
Synonym 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
PubChem CID 15731
ChEBI CHEBI:28970
SMILES OC1=CC=CC=C1C1=CC=CC=C1O

Specifications

Melting Point 107°C to 111°C
Density 1.34
Boiling Point 314°C to 316°C
Flash Point 154°C (309°F)
Quantity 500 g
Beilstein 1638363
Solubility Information Insoluble in water.
IUPAC Name [1,1'-biphenyl]-2,2'-diol
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2,2'-Dihydroxybiphenyl
Show More Show Less
Hazard Category H302-H315-H318
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-303-0
RTECSNumber DV4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only