missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dihydroxyazobenzene 98.0+%, TCI America™

Catalog No. p-7118252
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50051G 1 g
A50055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50051G Supplier TCI America Supplier No. A50051G

Chemical Identifiers

CAS 2050-14-8
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD00002182
InChI Key JLRUNUCYUPAYKT-UHFFFAOYSA-N
PubChem CID 6811824
IUPAC Name 6-[(2-hydroxyphenyl)hydrazinylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC=C(C(=C1)NN=C2C=CC=CC2=O)O

Specifications

Melting Point 175°C
Quantity 1 g
Synonym 2,2′C-Azodiphenol, DHAB
Formula Weight 214.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxyazobenzene [Spectrophotometric and fluorimetric reagent for Al, Mg and other met