missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorosuccinic acid, 94%

Catalog No. aal0077306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0077306 5 g
AAL0077303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0077306 Supplier Thermo Scientific Chemicals Supplier No. L0077306

CAS: 665-31-6 | C4H4F2O4 | 154.07 g/mol

2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.

Chemical Identifiers

CAS 665-31-6
Molecular Formula C4H4F2O4
Molecular Weight (g/mol) 154.07
MDL Number MFCD00040970
InChI Key ZYLFHISLYSHWRH-UHFFFAOYSA-N
Synonym 2,2-difluorosuccinic acid, 2,2-difluorosuccinicacid, butanedioic acid, 2,2-difluoro, pubchem23428, 2,2-difluoro-butanedioic acid, butanedioic acid,2,2-difluoro, 2,2-difluoro succinic acid, 2,2-difluorobutane-1,4-dioic acid
PubChem CID 69581
IUPAC Name 2,2-difluorobutanedioic acid
SMILES OC(=O)CC(F)(F)C(O)=O

Specifications

Melting Point 139°C to 144°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 1773355
Solubility Information Soluble in water.
Formula Weight 154.07
Percent Purity 94%
Chemical Name or Material 2,2-Difluorosuccinic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 211-558-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only