CAS RN 665-31-6
IUPAC Name:
2,2-difluorobutanedioic acid
Synonyms:
2,2-difluorobutanedioic acid
Molecular Weight (g/mol):
154.07
Molecular Formula:
C4H4F2O4
InChi Key:
ZYLFHISLYSHWRH-UHFFFAOYSA-N
SMILES:
OC(=O)CC(F)(F)C(O)=O
1
–
1
of
1
results
1
Promotions Available
2,2-Difluorosuccinic acid, 94%
2,2-Difluorosuccinic acid, 94%
CAS: 665-31-6 Molecular Formula: C4H4F2O4 Molecular Weight (g/mol): 154.07 MDL Number: MFCD00040970 InChI Key: ZYLFHISLYSHWRH-UHFFFAOYSA-N Synonym: 2,2-difluorosuccinic acid,2,2-difluorosuccinicacid,butanedioic acid, 2,2-difluoro,pubchem23428,2,2-difluoro-butanedioic acid,butanedioic acid,2,2-difluoro,2,2-difluoro succinic acid,2,2-difluorobutane-1,4-dioic acid PubChem CID: 69581 IUPAC Name: 2,2-difluorobutanedioic acid SMILES: OC(=O)CC(F)(F)C(O)=O