missing translation for 'onlineSavingsMsg'
Learn More

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%

CAS: 128073-33-6 | C5H6F2O2 | 136.098 g/mol

Supplier:  Thermo Scientific Chemicals H3305503

 View more versions of this product

Catalog No. AAH3305503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
62°C to 64°C
MFCD07777163
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
CC1(CC1(F)F)C(=O)O
136.098
136.1
128073-33-6
C5H6F2O2
1 g
HLFLYOQLHYYNLT-UHFFFAOYSA-N
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
14695229
97%

Chemical Identifiers

128073-33-6
136.098
HLFLYOQLHYYNLT-UHFFFAOYSA-N
14695229
CC1(CC1(F)F)C(=O)O
C5H6F2O2
MFCD07777163
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only