missing translation for 'onlineSavingsMsg'
Learn More

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%

Catalog No. p-7031836
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33055MD 250 mg
AAH3305503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33055MD Supplier Thermo Scientific Chemicals Supplier No. H33055MD
Only null left

CAS: 128073-33-6 | C5H6F2O2 | 136.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128073-33-6
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD07777163
InChI Key HLFLYOQLHYYNLT-UHFFFAOYSA-N
Synonym 2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
PubChem CID 14695229
IUPAC Name 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
SMILES CC1(CC1(F)F)C(=O)O

Specifications

Melting Point 62°C to 64°C
Quantity 250 mg
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material (±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only