Learn More
2,2-Difluoro-1,3-benzodioxole, 97%
CAS: 1583-59-1 | C7H4F2O2 | 158.10 g/mol
Supplier: Thermo Scientific Chemicals B2054006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,2-Difluoro-1,3-benzodioxole | |
| 1.308 | |
| 32°C (90°F) | |
| 1.443 | |
| 5 g | |
| 1307648 | |
| DGCOGZQDAXUUBY-UHFFFAOYSA-N | |
| 2,2-difluoro-2H-1,3-benzodioxole | |
| 74103 | |
| 97% |
| 1583-59-1 | |
| 129°C to 130°C | |
| C7H4F2O2 | |
| MFCD00236217 | |
| UN1993 | |
| 2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9d | |
| FC1(F)OC2=CC=CC=C2O1 | |
| 158.10 | |
| 158.11 |
Chemical Identifiers
| 1583-59-1 | |
| 158.10 | |
| DGCOGZQDAXUUBY-UHFFFAOYSA-N | |
| 74103 | |
| FC1(F)OC2=CC=CC=C2O1 |
| C7H4F2O2 | |
| MFCD00236217 | |
| 2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9d | |
| 2,2-difluoro-2H-1,3-benzodioxole |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 216-431-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only