accessibility menu, dialog, popup

UserName

FFOO

CAS RN 1583-59-1

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxole
Synonyms: 2,2-difluoro-1,3-benzodioxole
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: DGCOGZQDAXUUBY-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=CC=C2O1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)