missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichloropropionic Acid 95.0+%, TCI America™
$121.16 - $121.16
Chemical Identifiers
| CAS | 75-99-0 |
|---|---|
| Molecular Formula | C3H4Cl2O2 |
| Molecular Weight (g/mol) | 142.963 |
| MDL Number | MFCD00004178 |
| InChI Key | NDUPDOJHUQKPAG-UHFFFAOYSA-N |
| Synonym | 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae |
| PubChem CID | 6418 |
| ChEBI | CHEBI:81859 |
| IUPAC Name | 2,2-dichloropropanoic acid |
| SMILES | CC(C(=O)O)(Cl)Cl |
Chemical Identifiers
| 75-99-0 | |
| 142.963 | |
| NDUPDOJHUQKPAG-UHFFFAOYSA-N | |
| 6418 | |
| 2,2-dichloropropanoic acid |
| C3H4Cl2O2 | |
| MFCD00004178 | |
| 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae | |
| CHEBI:81859 | |
| CC(C(=O)O)(Cl)Cl |
Specifications
| 75-99-0 | |
| 99°C | |
| MFCD00004178 | |
| 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae | |
| CC(C(=O)O)(Cl)Cl | |
| 142.963 | |
| CHEBI:81859 | |
| ≥95.0% (GC) | |
| 2,2-Dichloropropionic Acid |
| Yellow | |
| C3H4Cl2O2 | |
| 1 g | |
| NDUPDOJHUQKPAG-UHFFFAOYSA-N | |
| 2,2-dichloropropanoic acid | |
| 6418 | |
| 142.96 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3913
RTECSNumber : UF0690000
TSCA : Yes