missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichlorobutane 88.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 104°C |
| Quantity | 10 mL |
| UN Number | 1993 |
| Formula Weight | 127.01 |
| Percent Purity | ≥88.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2-Dichlorobutane |
Safety and Handling
| EINECSNumber | (2)-0061 |
| TSCA | Yes |