accessibility menu, dialog, popup

UserName

H3CCH3ClCl

CAS RN 4279-22-5

IUPAC Name: 2,2-dichlorobutane
Synonyms: butane, 2,2-dichloro-
Molecular Weight (g/mol): 127.01
Molecular Formula: C4H8Cl2
InChi Key: BSRTYNDWQXVCKR-UHFFFAOYSA-N
SMILES: CCC(C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results