missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichloro-1,1-difluoroethyl methyl ether, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Dichloro-1,1-difluoroethyl methyl ether is used as a lipid soluble anesthetic agent. It is a neuromuscular blocking agent given concurrently to get the desired degree of muscular relaxation. Further, it is a powerful analgesic agent.
Solubility
Miscible with alcohol, acetone, chloroform, ether, fixed oils and benzene. Immiscible with water.
Notes
Incompatible with oxidizing agents.
2,2-Dichloro-1,1-difluoroethyl methyl ether is used as a lipid soluble anesthetic agent. It is a neuromuscular blocking agent given concurrently to get the desired degree of muscular relaxation. Further, it is a powerful analgesic agent.
Solubility
Miscible with alcohol, acetone, chloroform, ether, fixed oils and benzene. Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -36°C |
| Density | 1.44 |
| Boiling Point | 105°C |
| Flash Point | 37°C (98°F) |
| Quantity | 5 g |
| UN Number | UN3271 |
| Beilstein | 1737766 |
| Merck Index | 14,5994 |
| Solubility Information | Miscible with alcohol,acetone,chloroform,ether,fixed oils and benzene. Immiscible with water. |
| Formula Weight | 164.97 |
| Show More |
Safety and Handling
| Hazard Category | H226-H300-H319-H341 |
| Hazard Statement | GHS H Statement H226-H341-H319 Flammable liquid and vapor. Suspected of causing genetic defects. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P210-P233-P240-P241-P242-P243-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S. |
| EINECSNumber | 200-956-0 |
| RTECSNumber | KN7820000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only