accessibility menu, dialog, popup

UserName

H3COFFClCl

CAS RN 76-38-0

IUPAC Name: 2,2-dichloro-1,1-difluoro-1-methoxyethane
Synonyms: methoxyflurane methofane methoxiflurane analgizer C3H4cl2F2O ingalan metossiflurano anecotan methoflurane MOF
Molecular Weight (g/mol): 164.96
Molecular Formula: C3H4Cl2F2O
InChi Key: RFKMCNOHBTXSMU-UHFFFAOYSA-N
SMILES: COC(F)(F)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results