missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloro-1,1-difluoroethyl methyl ether, 97%, Thermo Scientific Chemicals

Catalog No. AAL1728506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1728506 5 g
AAL1728514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1728506 Supplier Thermo Scientific Chemicals Supplier No. L1728506
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dichloro-1,1-difluoroethyl methyl ether is used as a lipid soluble anesthetic agent. It is a neuromuscular blocking agent given concurrently to get the desired degree of muscular relaxation. Further, it is a powerful analgesic agent.

Solubility
Miscible with alcohol, acetone, chloroform, ether, fixed oils and benzene. Immiscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 76-38-0
Molecular Formula C3H4Cl2F2O
Molecular Weight (g/mol) 164.96
MDL Number MFCD00040144
InChI Key RFKMCNOHBTXSMU-UHFFFAOYSA-N
Synonym methoxyflurane, methoxyfluran, penthrane, methoflurane, metoxifluran, metoxfluran, methoxane, anecotan, metofane, pentrane
PubChem CID 4116
ChEBI CHEBI:6843
IUPAC Name 2,2-dichloro-1,1-difluoro-1-methoxyethane
SMILES COC(F)(F)C(Cl)Cl

Specifications

Melting Point -36°C
Density 1.44
Boiling Point 105°C
Flash Point 37°C (98°F)
Quantity 5 g
UN Number UN3271
Beilstein 1737766
Merck Index 14,5994
Solubility Information Miscible with alcohol,acetone,chloroform,ether,fixed oils and benzene. Immiscible with water.
Formula Weight 164.97
Percent Purity 97%
Chemical Name or Material 2,2-Dichloro-1,1-difluoroethyl methyl ether
Show More Show Less
Hazard Category H226-H300-H319-H341
Hazard Statement GHS H Statement
H226-H341-H319
Flammable liquid and vapor.
Suspected of causing genetic defects.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 200-956-0
RTECSNumber KN7820000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only