missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dibromobiphenyl, 98%

CAS: 13029-09-9 | C12H8Br2 | 312.01 g/mol

$105.37 - $105.37

Chemical Identifiers

CAS 13029-09-9
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.01
MDL Number MFCD00093707
InChI Key DRKHIWKXLZCAKP-UHFFFAOYSA-N
Synonym 2,2'-dibromobiphenyl, 2,2'-dibromo-1,1'-biphenyl, o,o'-dibromobiphenyl, 1-bromo-2-2-bromophenyl benzene, 1,1'-biphenyl, 2,2'-dibromo, unii-gsl06p6p2l, biphenyl, 2,2'-dibromo, gsl06p6p2l, 1,1'-biphenyl, 2,2'-dibromo-9ci, biphenyl,2'-dibromo
PubChem CID 83060
IUPAC Name 1-bromo-2-(2-bromophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC355930010
SDS View Documents
Thermo Scientific Chemicals
355930010
1 g Glass bottle
Each for $105.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13029-09-9
312.01
DRKHIWKXLZCAKP-UHFFFAOYSA-N
83060
C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br
C12H8Br2
MFCD00093707
2,2'-dibromobiphenyl, 2,2'-dibromo-1,1'-biphenyl, o,o'-dibromobiphenyl, 1-bromo-2-2-bromophenyl benzene, 1,1'-biphenyl, 2,2'-dibromo, unii-gsl06p6p2l, biphenyl, 2,2'-dibromo, gsl06p6p2l, 1,1'-biphenyl, 2,2'-dibromo-9ci, biphenyl,2'-dibromo
1-bromo-2-(2-bromophenyl)benzene

Specifications

13029-09-9
100.0
White
97.5% min. (GC)
C12H8Br2
1 g
DRKHIWKXLZCAKP-UHFFFAOYSA-N
1-bromo-2-(2-bromophenyl)benzene
83060
98%
2, 2'-Dibromobiphenyl
97.5
79°C to 83°C
Authentic
Glass bottle
MFCD00093707
2,2'-dibromobiphenyl, 2,2'-dibromo-1,1'-biphenyl, o,o'-dibromobiphenyl, 1-bromo-2-2-bromophenyl benzene, 1,1'-biphenyl, 2,2'-dibromo, unii-gsl06p6p2l, biphenyl, 2,2'-dibromo, gsl06p6p2l, 1,1'-biphenyl, 2,2'-dibromo-9ci, biphenyl,2'-dibromo
C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br
312.01
312.01
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only