missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dibromobiphenyl, 98%

Catalog No. aaa1763203
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
AAA1763203 1 g
AAA1763206 5 g
AAA1763214 25 g
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Catalog No. AAA1763203

Supplier: Thermo Scientific Chemicals A1763203

Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 13029-09-9 | C12H8Br2 | 312.004 g/mol

2,2'-Dibromobiphenyl is used to produce 5,5-dimethyl-5H-dibenzosilole at the temperature of -78 - 20°C. It will need reagent n-BuLi and solvents diethyl ether, hexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dibromobiphenyl is used to produce 5,5-dimethyl-5H-dibenzosilole at the temperature of -78 - 20°C. It will need reagent n-BuLi and solvents diethyl ether, hexane.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 13029-09-9
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.004
MDL Number MFCD00093707
InChI Key DRKHIWKXLZCAKP-UHFFFAOYSA-N
Synonym 2,2'-dibromobiphenyl, 2,2'-dibromo-1,1'-biphenyl, o,o'-dibromobiphenyl, 1-bromo-2-2-bromophenyl benzene, 1,1'-biphenyl, 2,2'-dibromo, unii-gsl06p6p2l, biphenyl, 2,2'-dibromo, gsl06p6p2l, 1,1'-biphenyl, 2,2'-dibromo-9ci, biphenyl,2'-dibromo
PubChem CID 83060
IUPAC Name 1-bromo-2-(2-bromophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br

Specifications

Melting Point 78°C to 81°C
Quantity 1 g
UN Number UN3152
Solubility Information Insoluble in water.
Formula Weight 312.02
Percent Purity 98%
Chemical Name or Material 2,2'-Dibromobiphenyl
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only