missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromo-4'-fluoroacetophenone, 98%

Catalog No. aal1978803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1978803 1 g
AAL1978806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1978803 Supplier Thermo Scientific Chemicals Supplier No. L1978803
Only null left

CAS: 7542-64-5 | C8H5Br2FO | 295.933 g/mol

2,2-Dibromo-4'-fluoroacetophenone is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dibromo-4′-fluoroacetophenone is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 7542-64-5
Molecular Formula C8H5Br2FO
Molecular Weight (g/mol) 295.933
MDL Number MFCD03791309
InChI Key XFFGHBAVJPOWDG-UHFFFAOYSA-N
Synonym 2,2-dibromo-1-4-fluorophenyl ethanone, alpha,alpha-dibromo-4-fluoroacetophenone, 2,2-dibromo-4'-fluoroacetophenone, 4'-fluoro-alpha,alpha-dibromoacetophenone, 2,2-dibromo-1-4-fluorophenyl ethan-1-one, 2,2-bis bromanyl-1-4-fluorophenyl ethanone
PubChem CID 2783255
IUPAC Name 2,2-dibromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)C(Br)Br)F

Specifications

Melting Point 36°C to 38°C
Quantity 1 g
UN Number UN3261
Beilstein 2256317
Solubility Information Insoluble in water.
Formula Weight 295.93
Percent Purity 98%
Chemical Name or Material 2,2-Dibromo-4'-fluoroacetophenone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only