missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Diaminobiphenyl 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 78°C |
| Boiling Point | 151°C |
| Quantity | 1 g |
| Formula Weight | 184.24 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Diaminobiphenyl |
Safety and Handling
| RTECSNumber | DV2099000 |
| TSCA | No |