missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, 99%

Catalog No. AAA1307430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1307430 250 g
AAA1307418 50 g
AAA130740B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1307430 Supplier Thermo Scientific Chemicals Supplier No. A1307430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 628-89-7 | C4H9ClO2 | 124.56 g/mol

2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

Solubility
Miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 628-89-7
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00002870
InChI Key LECMBPWEOVZHKN-UHFFFAOYSA-N
Synonym 2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
PubChem CID 12361
IUPAC Name 2-(2-chloroethoxy)ethanol
SMILES OCCOCCCl

Specifications

Density 1.18
Boiling Point 185°C
Flash Point 90°C (194°F)
Refractive Index 1.4535
Linear Formula ClCH2CH2OCH2CH2OH
Quantity 250 g
Beilstein 506015
Solubility Information Miscible with water.
Formula Weight 124.57
Percent Purity 99%
Chemical Name or Material 2-(2-Chloroethoxy)ethanol
Show More Show Less
Hazard Category H227-H315-H318
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 211-059-9
RTECSNumber KK1350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only