accessibility menu, dialog, popup

UserName

HOOCl

CAS RN 628-89-7

IUPAC Name: 2-(2-chloroethoxy)ethan-1-ol
Synonyms: ethanol, 2-(2-chloroethoxy)-
Molecular Weight (g/mol): 124.56
Molecular Formula: C4H9ClO2
InChi Key: LECMBPWEOVZHKN-UHFFFAOYSA-N
SMILES: OCCOCCCl
Boiling Point 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)