missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7124152
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5663200MG 200 mg
B56631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5663200MG Supplier TCI America Supplier No. B5663200MG
Only null left

Chemical Identifiers

CAS 77989-15-2
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.268
MDL Number MFCD29047055
InChI Key MSTJGWCHJCZPEQ-UHFFFAOYSA-N
PubChem CID 12689834
IUPAC Name 2-(2-bromophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=CC=C3Br)C4=CC=CC=C4

Specifications

Melting Point 144°C
Color White
Quantity 200 mg
Formula Weight 388.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine