missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™

Supplier:  TCI America B36531G

 View more versions of this product

Catalog No. B36531G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine
C16H12BBrN2
1 g
VHWWJBPYVUEFBM-UHFFFAOYSA-N
3-(2-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
53384411
≥98.0% (GC,N)
927384-42-7
MFCD16038144
2-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
BrC1=CC=CC=C1B1NC2=CC=CC3=CC=CC(N1)=C23
323.00
323.00
Crystalline Powder

Chemical Identifiers

927384-42-7
323.00
VHWWJBPYVUEFBM-UHFFFAOYSA-N
53384411
BrC1=CC=CC=C1B1NC2=CC=CC3=CC=CC(N1)=C23
C16H12BBrN2
MFCD16038144
2-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
3-(2-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions