missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)-1,3-dioxolane, 98%

Catalog No. AC393240250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393240250 25 g Glass bottle
AC393240050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393240250 Supplier Thermo Scientific Chemicals Supplier No. 393240250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34824-58-3 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34824-58-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00155124
InChI Key IWSGKSUCFVOWQU-UHFFFAOYSA-N
Synonym 2-2-bromophenyl-1,3-dioxolane, 2-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-2-bromophenyl, 1-bromo-2-1,3-dioxolan-2-yl benzene, 2-bromophenyldioxolane, 2-bromo benzaldehyde ethylene glycol acetal, 2-2-bromo-phenyl-1,3 dioxolane, pubchem3729, acmc-1csyy, bromobenzaldehyde ethylene acetal
PubChem CID 553534
IUPAC Name 2-(2-bromophenyl)-1,3-dioxolane
SMILES BrC1=CC=CC=C1C1OCCO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5690 (20°C, 589nm)
Quantity 25 g
Formula Weight 229.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Bromophenyl)-1, 3-dioxolane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only