missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)pyridine 98.0+%, TCI America™

Catalog No. p-7123963
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51131G 1 g
B51135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51131G Supplier TCI America Supplier No. B51131G

Chemical Identifiers

CAS 4373-60-8
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD09749846
InChI Key WLPFTJXVEBANAM-UHFFFAOYSA-N
Synonym 2-3-bromophenyl pyridine, pyridine, 2-3-bromophenyl, 2-3-bromo-phenyl-pyridine, 2-3'-bromophenyl pyridine, 3-2-pyridinyl phenyl bromide, 3,5-pyrid-2-yl bromobenzene
PubChem CID 11115506
IUPAC Name 2-(3-bromophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC=C2)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromophenyl)pyridine