missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-2-methyl-1,3-dioxolane 96.0+%, TCI America™

Catalog No. p-7119143
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15985G 5 g
B159825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15985G Supplier TCI America Supplier No. B15985G
Only null left

Chemical Identifiers

CAS 37865-96-6
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00191321
InChI Key SZLPRRVCSGZARF-UHFFFAOYSA-N
PubChem CID 11805619
IUPAC Name 2-(2-bromoethyl)-2-methyl-1,3-dioxolane
SMILES CC1(OCCO1)CCBr

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator