missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-1,3-dioxolane, 95%, stab. with silver

Catalog No. p-7081456
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1144109 10 g
AAA1144118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1144109 Supplier Thermo Scientific Chemicals Supplier No. A1144109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18742-02-4 | C5H9BrO2 | 181.03 g/mol

2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.

Solubility
It is immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive. Store under inert gas.

Chemical Identifiers

CAS 18742-02-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00003216
InChI Key GGZQLTVZPOGLCC-UHFFFAOYSA-N
Synonym 2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
PubChem CID 87776
IUPAC Name 2-(2-bromoethyl)-1,3-dioxolane
SMILES BrCCC1OCCO1

Specifications

Density 1.512
Boiling Point 68°C to 70°C (8 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.479
Quantity 10 g
UN Number UN2810
Beilstein 103516
Sensitivity Light sensitive
Solubility Information It is immiscible with water.
Formula Weight 181.04
Percent Purity 95%
Chemical Name or Material 2- (2-Bromoethyl)-1,3-dioxolane, Stabilized with silver
Show More Show Less
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 242-551-1
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only