accessibility menu, dialog, popup

UserName

BrOO

CAS RN 18742-02-4

IUPAC Name: 2-(2-bromoethyl)-1,3-dioxolane
Synonyms: 2-(2-bromoethyl)-1,3-dioxolane
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: GGZQLTVZPOGLCC-UHFFFAOYSA-N
SMILES: BrCCC1OCCO1
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)