missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-1,3-dioxolane 90.0+%, TCI America™

Catalog No. B113225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B113225G 25 g
B1132100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B113225G Supplier TCI America Supplier No. B113225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18742-02-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00003216
InChI Key GGZQLTVZPOGLCC-UHFFFAOYSA-N
Synonym 2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
PubChem CID 87776
IUPAC Name 2-(2-bromoethyl)-1,3-dioxolane
SMILES BrCCC1OCCO1

Specifications

Color Yellow
Boiling Point 73°C
Quantity 25 g
Formula Weight 181.03
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethyl)-1,3-dioxolane
TSCA No
Recommended Storage Freezer