missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-8-hydroxyquinoline Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7117183
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10055G 5 g
A100525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10055G Supplier TCI America Supplier No. A10055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21302-43-2
Molecular Formula C9H10Cl2N2O
Molecular Weight (g/mol) 233.092
MDL Number MFCD00012737
InChI Key VTQDJAUGGZFPOI-UHFFFAOYSA-N
Synonym 5-Amino-8-quinolinol Dihydrochloride
PubChem CID 16211947
IUPAC Name 5-aminoquinolin-8-ol;dihydrochloride
SMILES C1=CC2=C(C=CC(=C2N=C1)O)N.Cl.Cl

Specifications

Quantity 5 g
Formula Weight 233.09
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-8-hydroxyquinoline Dihydrochloride