missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bis(trifluoromethyl)benzidine, 97%

Catalog No. AC457570050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC457570050 Supplier Thermo Scientific Chemicals Supplier No. 457570050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 341-58-2 | C14H10F6N2 | 320.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 341-58-2
Molecular Formula C14H10F6N2
Molecular Weight (g/mol) 320.24
MDL Number MFCD00190155
InChI Key NVKGJHAQGWCWDI-UHFFFAOYSA-N
Synonym 2,2'-bis trifluoromethyl benzidine, 2,2'-bis trifluoromethyl-1,1'-biphenyl-4,4'-diamine, 4-4-amino-2-trifluoromethyl phenyl-3-trifluoromethyl aniline, 2,2'-bis trifluoromethyl-4,4'-diaminobiphenyl, 2,2'-bis-trifluoromethyl-biphenyl-4,4'-diamine, tfmb, 2,2'-bis trifluoromethyl-4,4'-biphenyldiamine, 4,4'-diamino-2,2'-bis trifluoromethyl biphenyl, 1,1'-biphenyl-4,4'-diamine, 2,2'-bis trifluoromethyl, 4-4-azanyl-2-trifluoromethyl phenyl-3-trifluoromethyl aniline
PubChem CID 629349
IUPAC Name 4-[4-amino-2-(trifluoromethyl)phenyl]-3-(trifluoromethyl)aniline
SMILES NC1=CC(=C(C=C1)C1=C(C=C(N)C=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 183.0°C to 184.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 320.24
Percent Purity 97%
Chemical Name or Material 2,2'-Bis(trifluoromethyl)benzidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

RUO – Research Use Only