missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic Acid 97.0+%, TCI America™

Supplier:  TCI America B1192500G

 View more versions of this product

Catalog No. B1192500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,2-Bis(hydroxymethyl)propionic Acid
185°C
200°C
MFCD00004199
2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
CC(CO)(CO)C(=O)O
134.13
134.13
Crystalline Powder
4767-03-7
White
C5H10O4
500 g
PTBDIHRZYDMNKB-UHFFFAOYSA-N
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
78501
≥97.0% (T)

Chemical Identifiers

4767-03-7
134.13
PTBDIHRZYDMNKB-UHFFFAOYSA-N
78501
CC(CO)(CO)C(=O)O
C5H10O4
MFCD00004199
2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1328

RTECSNumber : UA2465265

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions