missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic Acid 97.0+%, TCI America™

Catalog No. p-7118909
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119225G 25 g
B1192500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B119225G Supplier TCI America Supplier No. B119225G
Only null left

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00004199
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N
Synonym 2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
PubChem CID 78501
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
SMILES CC(CO)(CO)C(=O)O

Specifications

Melting Point 185°C
Color White
Boiling Point 200°C
Quantity 25 g
Formula Weight 134.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(hydroxymethyl)propionic Acid
EINECSNumber (2)-1328
RTECSNumber UA2465265
TSCA Yes