missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(hydroxymethyl)butyric Acid 97.0+%, TCI America™
$67.11 - $357.90
Chemical Identifiers
| CAS | 10097-02-6 |
|---|---|
| Molecular Formula | C6H12O4 |
| Molecular Weight (g/mol) | 148.16 |
| MDL Number | MFCD00190088 |
| InChI Key | JVYDLYGCSIHCMR-UHFFFAOYSA-N |
| Synonym | 2,2-Dimethylolbutyric Acid |
| PubChem CID | 3301396 |
| IUPAC Name | 2,2-bis(hydroxymethyl)butanoic acid |
| SMILES | CCC(CO)(CO)C(O)=O |
Chemical Identifiers
| 10097-02-6 | |
| 148.16 | |
| JVYDLYGCSIHCMR-UHFFFAOYSA-N | |
| 3301396 | |
| CCC(CO)(CO)C(O)=O |
| C6H12O4 | |
| MFCD00190088 | |
| 2,2-Dimethylolbutyric Acid | |
| 2,2-bis(hydroxymethyl)butanoic acid |
Specifications
| 10097-02-6 | |
| White | |
| MFCD00190088 | |
| 2,2-Dimethylolbutyric Acid | |
| CCC(CO)(CO)C(O)=O | |
| 148.16 | |
| 148.16 | |
| Crystalline Powder |
| 112°C | |
| C6H12O4 | |
| 25 g | |
| JVYDLYGCSIHCMR-UHFFFAOYSA-N | |
| 2,2-bis(hydroxymethyl)butanoic acid | |
| 3301396 | |
| ≥97.0% (T) | |
| 2,2-Bis(hydroxymethyl)butyric Acid |
Safety and Handling
EINECSNumber : (2)-3817
TSCA : Yes