accessibility menu, dialog, popup

UserName

CH3OHOHOHO

CAS RN 10097-02-6

IUPAC Name: 2,2-bis(hydroxymethyl)butanoic acid
Synonyms: 2,2-bis(hydroxymethyl)butanoic acid
Molecular Weight (g/mol): 148.16
Molecular Formula: C6H12O4
InChi Key: JVYDLYGCSIHCMR-UHFFFAOYSA-N
SMILES: CCC(CO)(CO)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results