missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(hydroxymethyl)butyric Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 112°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 148.16 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2-Bis(hydroxymethyl)butyric Acid |
Safety and Handling
| EINECSNumber | (2)-3817 |
| TSCA | Yes |