missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Bis(diphenylphosphino)biphenyl 98.0+%, TCI America™
$61.61 - $350.38
Chemical Identifiers
| CAS | 84783-64-2 |
|---|---|
| Molecular Formula | C36H28P2 |
| Molecular Weight (g/mol) | 522.57 |
| MDL Number | MFCD03094574 |
| InChI Key | GRTJBNJOHNTQBO-UHFFFAOYSA-N |
| Synonym | 2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl |
| PubChem CID | 2734940 |
| IUPAC Name | [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane |
| SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
B2630100MG
|
TCI America
B2630100MG |
100 mg |
Each for $61.61
|
|
|||||
|
B26301G
|
TCI America
B26301G |
1 g |
Each for $350.38
|
|
|||||
Chemical Identifiers
| 84783-64-2 | |
| 522.57 | |
| GRTJBNJOHNTQBO-UHFFFAOYSA-N | |
| 2734940 | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C36H28P2 | |
| MFCD03094574 | |
| 2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl | |
| [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane |
Specifications
| 84783-64-2 | |
| White-Yellow | |
| MFCD03094574 | |
| 2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 522.57 | |
| 522.57 | |
| Crystalline Powder |
| 215°C | |
| C36H28P2 | |
| 100 mg | |
| GRTJBNJOHNTQBO-UHFFFAOYSA-N | |
| [2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane | |
| 2734940 | |
| ≥98.0% (GC) | |
| 2,2′-Bis(diphenylphosphino)biphenyl |
Safety and Handling
TSCA : No